Learn More
Hach Company Intellical™ LDO101 Laboratory Luminescent/Optical Dissolved Oxygen (DO) Sensor, 3 m Cable
Catalog No. 501997744 Shop All Hach Company Products
Encompass
Intellical™ LDO101 is a digital, luminescent/optical dissolved oxygen (LDO) probe. The laboratory version is available with a 1 or 3 meters cable. The LDO101 is fitted with an automatic pressure sensor module and a temperature sensor. The DO sensing cap comes with an iButton to track days in use and remind remaining life of the sensing cap element.The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-7744 Supplier Hach Company Supplier No. LDO10103
Intellical™ LDO101 is a digital, luminescent/optical dissolved oxygen (LDO) probe. The laboratory version is available with a 1 or 3 meters cable. The LDO101 is fitted with an automatic pressure sensor module and a temperature sensor. The DO sensing cap comes with an iButton to track days in use and remind remaining life of the sensing cap element.The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.