Learn More
Hach Company LDO sc Calibration Bag
Supplier:  Hach Company 5796600
Encompass
LDO sc Calibration Bag-6in x 16 Metalized PET bag with ziplock closure used during calibration of the Hach LDO Process Probe (Probe not included)
Catalog No. 50-002-06974