Learn More
Hach Company LDO2 Sen w/SC4500 2 Ch Kit
Catalog No. 5000207896 Shop All Hach Company Products
Encompass
LDO sc Model 2 Dissolved Oxygen Sensor-10 m Cable-Adapter-with SC4500-2 ChannelsKit including the LDO Model 2 Probe Mounting Conversion Adapter and SC4500 Controller (100-240 VAC) with 2 Channels For a supremely accurate DO measurements that does not require calibration and is membrane free turn to an innovator in DO technology and Hach s next generation LDO Probe
Catalog No. 50-002-07896 Supplier Hach Company Supplier No. LXV525.97KTO033
Add to Cart
Add to Cart
Request a Quote
LDO sc Model 2 Dissolved Oxygen Sensor-10 m Cable-Adapter-with SC4500-2 ChannelsKit including the LDO Model 2 Probe Mounting Conversion Adapter and SC4500 Controller (100-240 VAC) with 2 Channels For a supremely accurate DO measurements that does not require calibration and is membrane free turn to an innovator in DO technology and Hach s next generation LDO Probe