Learn More
Qorpak™ LDPE Cubitainer
Description
- Designed to collapse as product is dispense
- Non-glug design assures product flow in a continuous, uninterrupted stream
- Reversible well design permits opening to be raised or depressed for pouring and storage
- Can withstand rigors of hazardous chemical packaging
Cubitainer, Container
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (English) | 1 gal. |
| Capacity (Metric) | 4 L |
| Certifications/Compliance | UN Rated |
| Closure Color | White |
| Closure Liner | PE foam |
| Closure Material | Polypropylene |
| Closure Type | Screw-on |
| Color | Natural |
| Disposable | Single-use |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.