Learn More
Research Products International Corp 8 Ounce Bottle, LDPE, with 28 mm Cap, 12 per Package
SDP
Catalog No. 50488663
Encompass
Translucent flexible bottles with leakproof design and matched polypropylene closures offer greater visibility than HDPE bottles. Perfect for sampling and general storage of liquids, powders or viscous fluids. Bottles feature recycling code molded into base.
Catalog No. 50-488-663 Supplier Research Products International Corp Supplier No. F106210016
Translucent flexible bottles with leakproof design and matched polypropylene closures offer greater visibility than HDPE bottles. Perfect for sampling and general storage of liquids, powders or viscous fluids. Bottles feature recycling code molded into base.