Learn More
MSC Globe Scientific Round Squeeze Wash Bottle, LDPE, Wide Mouth, 250 mL, 5/Pk.
Catalog No. 5000514646
Encompass
These round squeeze wash bottles are designed for reliable everyday use in laboratory environments. Constructed from high-quality low-density polyethylene (LDPE), they offer excellent chemical resistance, durability, and flexibility. Each bottle is fitted with a one-piece polypropylene (PP) closure that maintains its shape after repeated use.
  • Manufactured from flexible and durable LDPE
  • Features a wide mouth opening for easy, spill-free filling
  • Includes a precision jet-tip aperture for controlled dispensing of water, solvents, and reagents
  • Designed with a square-shouldered profile
  • Equipped with a one-piece polypropylene closure
  • Suitable for rinsing glassware and dispensing cleaning solutions
Catalog No. 50-005-14646 Supplier MSC Supplier No. 60891413
These round squeeze wash bottles are designed for reliable everyday use in laboratory environments. Constructed from high-quality low-density polyethylene (LDPE), they offer excellent chemical resistance, durability, and flexibility. Each bottle is fitted with a one-piece polypropylene (PP) closure that maintains its shape after repeated use.
  • Manufactured from flexible and durable LDPE
  • Features a wide mouth opening for easy, spill-free filling
  • Includes a precision jet-tip aperture for controlled dispensing of water, solvents, and reagents
  • Designed with a square-shouldered profile
  • Equipped with a one-piece polypropylene closure
  • Suitable for rinsing glassware and dispensing cleaning solutions