Learn More
AVANTOR MASTERFLEX (VWR LLC) Low-Density Polyethylene (LDPE) Tubing, 0.17"" x 1/4"", 100 ft/bag
Catalog No. NC9354031
Encompass
Applications: General low pressure and temperature, instrumentation, corrosive atmosphere, and food processing applications. Characteristics: Broad chemical, solvent, and corrosive atmosphere resistance with good dimensional stability. Translucent. Certification: FDA-compliant (21 CFR 177.1520); meets USDA requirements Temperature range: ¿100 to 175¿F (¿73 to 80¿C) Sterilize: Ethylene oxide.
Catalog No. NC9354031 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9587603
Applications: General low pressure and temperature, instrumentation, corrosive atmosphere, and food processing applications. Characteristics: Broad chemical, solvent, and corrosive atmosphere resistance with good dimensional stability. Translucent. Certification: FDA-compliant (21 CFR 177.1520); meets USDA requirements Temperature range: ¿100 to 175¿F (¿73 to 80¿C) Sterilize: Ethylene oxide.