Learn More
AVANTOR MASTERFLEX (VWR LLC) Low-Density Polyethylene (LDPE) Tubing, 1/4"" x 3/8"", 100 ft/bag
Catalog No. NC0283375
Encompass
Applications: General low pressure and temperature, instrumentation, corrosive atmosphere, and food processing applications. Characteristics: Broad chemical, solvent, and corrosive atmosphere resistance with good dimensional stability. Translucent. Certification: FDA-compliant (21 CFR 177.1520); meets USDA requirements Temperature range: ¿100 to 175¿F (¿73 to 80¿C) Sterilize: Ethylene oxide.
Catalog No. NC0283375 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9587605
Applications: General low pressure and temperature, instrumentation, corrosive atmosphere, and food processing applications. Characteristics: Broad chemical, solvent, and corrosive atmosphere resistance with good dimensional stability. Translucent. Certification: FDA-compliant (21 CFR 177.1520); meets USDA requirements Temperature range: ¿100 to 175¿F (¿73 to 80¿C) Sterilize: Ethylene oxide.