Learn More
Sigma Aldrich Fine Chemicals Biosciences LDPE wash bottles Narrow-m
Catalog No. 502900483
Encompass
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.
Catalog No. 50-290-0483 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z10464712EA
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.