Learn More
ALPAQUA ENGINEERING LLC LE MAGNET PLATE
Catalog No. NC0665458
Encompass
The Low Elution Magnet Plate from Alpaqua contains a unique plate docking structure that enables nucleic acid elution in as little as 8ul. It is designed for use with a variety of 96-well PCR plates.
Catalog No. NC0665458 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000350
The Low Elution Magnet Plate from Alpaqua contains a unique plate docking structure that enables nucleic acid elution in as little as 8ul. It is designed for use with a variety of 96-well PCR plates.