Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Series Generation 2 Ultra Low Temp Freezers Foundational Service eLearning Course

Catalog No. 01670095 Shop All Thermo Scientific Products
Change view
Click to view available options
Duration:
30 to 60 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration
01-670-095 30 to 60 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-095 Supplier Thermo Scientific™ Supplier No. TULTTSXG2F
Add to Cart
Add to Cart

Flexible, self-paced, effective option to train and certify your operators on Thermo Fisher equipment. These courses are designed and executed by certified and qualified expert trainers; covers basic installation, startup, operation and user maintenance.

Thermo Scientific™ TSX Series Generation 2 Ultra Low Temp Freezer Foundation eLearning course provides you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning modules which cover different topics such as introduction and/or basic operations and/or user maintenance.

Thermo Scientific™ TSX Series Generation 2 Ultra Low Temp Freezer eLearning courses are designed to help you get the most out of your technology, no matter your skill level. Included in this course:

Getting Started

  • Basic setup and configuration
  • Navigating your device
  • Connecting the Wi-Fi dongle and Light bar

 

Core Features

  • Utilizing key functions
  • Customizing settings
  • Loading samples
  • Organizing the unit

 

User Maintenance

  • Defrosting the unit
  • Maintaining your device

 

eLearning Courses

Empowering You with Knowledge. Begin your journey to becoming a device expert! Utilize course certificates as a part of your compliance.

TRUSTED_SUSTAINABILITY

Specifications

Course Topic Thermo Scientific™ TSX Series Generation 2 Ultra Low Temp Freezer Foundation eLearning course
Access Availablity 1 year upon end-user first access to the course
Course Level Foundation
Duration 30 to 60 min.
For Use With (Equipment) TSX Series Generation 2 Ultra Low Temp Freezer Foundation eLearning course
Product Type Training
Product Line Thermo Scientific
Training Location Digital access through Online Learning Management System
Format Self-paced learning
Target Audience New user seeking training on instrument operation and basic experimentation
Languages English
Time and Date Digital access through Online Learning Management System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.