Learn More
Medchemexpress LLC Leachianone A | 97938-31-3 | 1 ML
SDP
Catalog No. 5000421233
Encompass_Preferred
Leachianone A, isolated from Radix Sophorae, demonstrates anti-malarial, anti-inflammatory, and cytotoxic properties. It induces apoptosis through both extrinsic and intrinsic pathways.
  • Induces apoptosis via both extrinsic and intrinsic pathways.
  • Exhibits marked inhibition on the survival of HepG2 cells.
  • Decreases the precursor of caspase-3.
  • Reduces protein levels of pro-forms of upstream initiator caspases.
  • Significantly diminishes tumor volume in animal models.
  • Soluble in DMSO at 100 mg/mL.
Catalog No. 50-004-21233 Supplier Medchemexpress LLC Supplier No. HYN228110MM/1ML
Leachianone A, isolated from Radix Sophorae, demonstrates anti-malarial, anti-inflammatory, and cytotoxic properties. It induces apoptosis through both extrinsic and intrinsic pathways.
  • Induces apoptosis via both extrinsic and intrinsic pathways.
  • Exhibits marked inhibition on the survival of HepG2 cells.
  • Decreases the precursor of caspase-3.
  • Reduces protein levels of pro-forms of upstream initiator caspases.
  • Significantly diminishes tumor volume in animal models.
  • Soluble in DMSO at 100 mg/mL.