Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, 10% (w/v) with 1% (v/v) Acetic Acid, Ricca Chemical
SDP

Catalog No. R42935001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R42935001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Exact match for part number R42935001A
Catalog No. R42935001A Supplier Ricca Chemical Company Supplier No. R42935001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Exact match for part number R42935001A

Chemical Identifiers

CAS 6080-56-4 , 7732-18-5 , 64-19-7
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Min % 8.83, 88.0, 0.93
CAS Max % 9.19, 92.0, 0.96
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution with 1% (v/v) Acetic Acid
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Lead Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.