Learn More
Key Scientific Products Lead Acetate Strips, 30 strips/pack
SDP
Catalog No. NC9506930
Encompass_Preferred
Used to detect the production of hydrogen sulfide by Enterobacteriaciae. The strips are more sensitive than TSI or KIA slants, and may show positive results when the agar butt reaction is inconclusive. Includes 30 tests per jar.
Catalog No. NC9506930 Supplier Key Scientific Products Supplier No. K375
Used to detect the production of hydrogen sulfide by Enterobacteriaciae. The strips are more sensitive than TSI or KIA slants, and may show positive results when the agar butt reaction is inconclusive. Includes 30 tests per jar.