Learn More
Electron Microscopy Sciences Lead Acetate, Trihydrate Reagent A.C.S. 100 GR

Supplier: Electron Microscopy Sciences 17600
CAS #6080-56-4 Pb(CH₃COO)2·₃H₂₀ F.W. 379.33
Assay: 99.0-103.0%
Lead Acetate is available in 25 ml bottles to avoid its classification as a hazardous chemical for shipping purposes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.