Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate Trihydrate (Certified ACS), Fisher Chemical™

Catalog No. L33250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
L33-250 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L33-250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ²-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75°C
Color White
pH 5.5 to 6.5
Physical Form Solid
Quantity 250 g
Assay Percent Range 99 to 103%
Linear Formula Pb(C2H3O2)2·3H2O
UN Number 1616
Formula Weight 379.3
Percent Purity 99 to 103%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Decomposition Information Decomposes information: >100°C
Chemical Name or Material Lead Acetate Trihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.