Learn More
Lead Acetate, Trihydrate, Crystal, Reagent, ACS, 99-103%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| Physical Form | Crystals or Needles |
| CAS Max % | 100% |
| Quantity | 50 kg |
| Packaging | Poly Bottle |
| UN Number | UN1616 |
| Percent Purity | 99 to 103% |
| Grade | ACS/Reagent |
| Chemical Name or Material | Lead Acetate, Trihydrate, Crystal |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.