Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, Trihydrate, Crystal, Technical, Spectrum™ Chemical
SDP

Catalog No. 18603760 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-760 50 kg Poly Bottle
18-603-757 500 g Poly Bottle
18-603-758 2.5 kg Poly Bottle
18-603-759 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-760 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L104150KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L1041, 6080-56-4, C4H6O4Pb·3H2O

Spectrum™ Chemical Lead Acetate, Trihydrate, Crystal, Technical , also known as lead diacetate, is used in men′s hair coloring products and as an astringent. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystals and/or Chunks
Quantity 50 kg
UN Number UN1616
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Lead Acetate, Trihydrate, Crystal
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.