Learn More
Research Products International Corp Lead Apron, Adjust-A-Fit, 24 Inches Wide, 36 Inches Long, Light Blue
SDP
Catalog No. 50213077
Encompass
Features two hook & loop fastening flaps at the waist, which aid in relieving weight off the shoulders and includes adjustable padded shoulders. Its comfortable fit is attained by varying the position of the flaps. Shoulder pads included.
This item is not returnable. View return policy
Catalog No. 50-213-077 Supplier Research Products International Corp Supplier No. 103538LB
Features two hook & loop fastening flaps at the waist, which aid in relieving weight off the shoulders and includes adjustable padded shoulders. Its comfortable fit is attained by varying the position of the flaps. Shoulder pads included.