Learn More
Research Products International Corp Lead Apron, Adjust-A-Fit, 24 Inches Wide, 32 Inches Long, Beige
SDP
Catalog No. 50213070
Encompass
Features hook & loop closure at the waist, which aid in relieving weight off the shoulders. Its comfortable fit is attained by varying the position of the flaps. Shoulder pads included.
This item is not returnable. View return policy
Catalog No. 50-213-070 Supplier Research Products International Corp Supplier No. 103532
Features hook & loop closure at the waist, which aid in relieving weight off the shoulders. Its comfortable fit is attained by varying the position of the flaps. Shoulder pads included.