Learn More
Wolf X-Ray Corporation X-ray Protective Apron, Easy Wrap, Large, Light Weight Lead, with Thyroid Collar, 0.5mm, 24" x 42", Medium, Blue
SDP
Catalog No. NC1319816
Encompass_Preferred
Lightweight Lead. The two side closures allow you to easily custom-fit the weight distribution comfortably on your shoulders. 0.5 mm protection
This item is not returnable. View return policy
Catalog No. NC1319816 Supplier Wolf X-Ray Corporation Supplier No. 65023TC16
Lightweight Lead. The two side closures allow you to easily custom-fit the weight distribution comfortably on your shoulders. 0.5 mm protection