Learn More
Vee Gee Heathrow Scientific Lead C Ring 250-1000mL flasks
Catalog No. 502886375
Encompass_Preferred
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.* Vinyl-coated lead ring will not mark or scratch benchtops and labware* C-Shape Style allows some manual size variation for a customized fit
Catalog No. 50-288-6375 Supplier Vee Gee Supplier No. HS8881B
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.* Vinyl-coated lead ring will not mark or scratch benchtops and labware* C-Shape Style allows some manual size variation for a customized fit