Learn More
Vee Gee Heathrow Scientific Lead C Ring 500-2000mL flasks
Catalog No. 502886383
Encompass_Preferred
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.* Vinyl-coated lead ring will not mark or scratch benchtops and labware* C-Shape Style allows some manual size variation for a customized fit
Catalog No. 50-288-6383 Supplier Vee Gee Supplier No. HS8881C
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.* Vinyl-coated lead ring will not mark or scratch benchtops and labware* C-Shape Style allows some manual size variation for a customized fit