Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Carbonate, ACS Reagent, Reagents

Catalog No. 01337925
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-925 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-925 Supplier Reagents Holdings Llc Supplier No. C1227000250A6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lead Carbonate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym Lead(II) Carbonate, Plumbous Carbonate
IUPAC Name λ²-lead(2+) carbonate
SMILES [Pb++].[O-]C([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 250 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 6.61
Chemical Name or Material Lead Carbonate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.