Learn More
Electron Microscopy Sciences Lead Citrate, Trihydrate 100 GR
SDP
Supplier:  Electron Microscopy Sciences 17810
Encompass_Preferred
CAS #512-26-5 Pb(C₆H₅O₇)2·₃H₂O F.W. 1053.82
References: The most widely used metal stain for ultra thin sections. Reynolds, E.S., J. Cell Bio., 17, 208 (1963) Sato, T., J. Electron Micro., 116, 133 (1976) Venable, J.H. and Coggeshall, R. (1965). A simplified lead citrate stain for use in electron microscopy. J. Cell Biol., 25:407 Famy, A. (1967). An extemporaneous lead citrate for electron microscopy. Proc. 25th Ann. EMSA Meet., p.148. Claitor's Pub. Division, Baton Rouge, LA.
Catalog No. 50-268-91