Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead citrate, trihydrate, 96%, Thermo Scientific™

Catalog No. AC413065000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413065000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC413065000 Supplier Thermo Scientific Chemicals Supplier No. 413065000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6107-83-1
Molecular Formula C12H16O17Pb3
Molecular Weight (g/mol) 1053.843
MDL Number MFCD00150456
InChI Key GMPMGSCJCDAUMP-UHFFFAOYSA-H
Synonym lead ii citrate trihydrate, acmc-20akng, citric acid lead ii salt trihydrate, tris ? 2-lead 2+ dicitrate trihydrate, 2-hydroxypropane-1,2,3-tricarboxylate; lead 2+ ; trihydrate, 1,2,3-propanetricarboxylicacid, 2-hydroxy-, lead 2+ salt, hydrate 2:3:3
PubChem CID 2735068
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;lead(2+);trihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Pb+2].[Pb+2].[Pb+2]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (HOC(COO-)(CH2COO-)2)2Pb3·3H2O
Quantity 500 g
Formula Weight 1053.85
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Lead citrate, trihydrate, 96%
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if inhaled. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. May cause damage to organs through prolonged or repeated exposure. May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. Wash face, hands and any exposed skin thoroughly after handling. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 208-141-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.