Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Cyanate, MP Biomedicals

Catalog No. ICN223310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN223310 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN223310 Supplier MP Biomedicals, Inc Supplier No. 0522331080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 592-05-2
Molecular Formula C2N2Pb
Molecular Weight (g/mol) 259.236
InChI Key UMTFPTCPRUEQHM-UHFFFAOYSA-N
Synonym lead cyanide, lead dicyanide, lead ii cyanide, plumbanedicarbonitrile, cyanure de plomb french, c.i. pigment yellow 48, lead cyanide pb cn 2, cyanide lead, cyanure de plomb
PubChem CID 61139
IUPAC Name dicyanolead
SMILES C(#N)[Pb]C#N

Specifications

Quantity 100 g
Formula Weight 291.24
Chemical Name or Material Lead Cyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.