Learn More
Andwin Scientific LEAD DONUT VINYL COATED 237G
SDP
Catalog No. NC9334737
Encompass_Preferred
The lead ring is vinyl-coated and in a shape of a donut. It can work with flasks with volume capacity of 125 to 500 mL. This product allows flasks to be immersed in water baths, lessening risks of breakage and spills. The lead ring weighs 237 grams.
Catalog No. NC9334737 Supplier Andwin Scientific Supplier No. EF8839A
The lead ring is vinyl-coated and in a shape of a donut. It can work with flasks with volume capacity of 125 to 500 mL. This product allows flasks to be immersed in water baths, lessening risks of breakage and spills. The lead ring weighs 237 grams.