Learn More
Sigma Aldrich Fine Chemicals Biosciences Lead foil, 300x300mm, thickness 1.0mm, as rolled, 99.95% | 7439-92-1 | MFCD00134050 | 1EA
Catalog No. 501771547
Encompass
Lead foil, 300x300mm, thickness 1.0mm, as rolled, 99.95% | Purity: 99.95% | Mol Wt: 207.2 | 7439-92-1 | MFCD00134050 | 1EA
This item is not returnable. View return policy
Catalog No. 50-177-1547 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF788287551EA
Lead foil, 300x300mm, thickness 1.0mm, as rolled, 99.95% | Purity: 99.95% | Mol Wt: 207.2 | 7439-92-1 | MFCD00134050 | 1EA