Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(Ii) Acetate Trihydrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6000888 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-88 250 g
60-008-86 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-008-88 Supplier Solstice Supplier No. 32307250G

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia, 99.5-102.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 250 g
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number 1616
Beilstein 3730298
Formula Weight 379.33g/mol
Percent Purity 99.5 to 102%
Grade ACS/puriss. p.a.
Chemical Name or Material Lead(Ii) Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P201-P273-P308 + P313-P501
Precautionary Statement H360Df-H373-H410
EINECSNumber 206-104-4
RTECSNumber OF8050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.