Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) carbonate, ACS reagent

Catalog No. p-4440135
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC212511000 100 g
AC212515000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC212511000 Supplier Thermo Scientific Chemicals Supplier No. 212511000
Only null left
Add to Cart

CAS: 598-63-0 | CO3Pb | 267.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O

Specifications

Melting Point 400.0°C
Quantity 100 g
Linear Formula PbCO3
Merck Index 10,1006
Solubility Information Solubility in water: insoluble
IUPAC Name λ²-lead(2+) carbonate
Formula Weight 267.2
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Lead(II) carbonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 209-943-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.