Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) citrate trihydrate, 99%, Thermo Scientific Chemicals

Catalog No. AAA1070122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1070122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1070122 Supplier Thermo Scientific Chemicals Supplier No. A1070122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6107-83-1
Molecular Formula C12H16O17Pb3
Quantity 100 g
MDL Number MFCD00150456
UN Number UN2291
Synonym lead ii citrate trihydrate, acmc-20akng, citric acid lead ii salt trihydrate, tris ? 2-lead 2+ dicitrate trihydrate, 2-hydroxypropane-1,2,3-tricarboxylate; lead 2+ ; trihydrate, 1,2,3-propanetricarboxylicacid, 2-hydroxy-, lead 2+ salt, hydrate 2:3:3
Solubility Information Soluble in cold water
InChI Key GMPMGSCJCDAUMP-UHFFFAOYSA-H
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Pb+2].[Pb+2].[Pb+2]
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;lead(2+);trihydrate
Molecular Weight (g/mol) 1053.843
PubChem CID 2735068
Formula Weight 1053.82 (999.77 Anhydrous)
Percent Purity 99%
Chemical Name or Material Lead (II) citrate trihydrate
Show More Show Less
Hazard Category H301-H330-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 208-141-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.