Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) formate, 90+%, Thermo Scientific™

Catalog No. AAA1435730
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1435730 250 g
AAA143570B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1435730 Supplier Thermo Scientific Chemicals Supplier No. A1435730

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-54-1
Molecular Formula C2H2O4Pb
Molecular Weight (g/mol) 297.234
MDL Number MFCD00050834
InChI Key ZGVSCHRFRACEIO-UHFFFAOYSA-L
Synonym lead diformate, lead formate, formic acid, lead 2+ salt, unii-2kjp65ymf4, lead ii formate, lead 2+ formate, 2kjp65ymf4, lead formate pb o2ch 2, lead formate pb hco2 2, formic acid, lead 2+ salt 2:1
PubChem CID 61216
IUPAC Name lead(2+);diformate
SMILES C(=O)[O-].C(=O)[O-].[Pb+2]

Specifications

Melting Point 190°C (decomposition)
Quantity 250 g
UN Number UN2291
Merck Index 14,5409
Formula Weight 297.23
Percent Purity ≥90%
Density 4.63
Chemical Name or Material Lead(II) formate
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 212-371-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.