Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) niobium(V) 2-ethylhexanoate decaethoxide, 99% (metals basis), 10% w/v in ethanol, Thermo Scientific™

Catalog No. AA36577AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36577AE 100 mL
AA36577AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36577AE Supplier Thermo Scientific Chemicals Supplier No. 36577AE

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00171335
Synonym Lead(II) niobium(V) 2-ethylhexano-ethoxide

Specifications

Quantity 100 mL
UN Number UN1170
Sensitivity Moisture Sensitive
Formula Weight 1130.04
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lead(II) niobium(V) 2-ethylhexanoate decaethoxide
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H311-H371-H332-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
May cause damage to organs.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.