Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lead(II) Perchlorate Hydrate

Catalog No. 45910025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-910-025G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 45-910-025G Supplier Sigma Aldrich Supplier No. 45910025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lead(II) perchlorate hydrate [Pb(ClO4)2. H2O] can be used as a precursor to synthesize: Pb(II) hydroxycarbonate (plumbonacrite) nanoparticles. Lead sulfide semiconductor nanoparticles by radiolytic method Lead coordination polymers.

Specifications

CAS 207500-00-3
Melting Point 177°C to 180°C
Molecular Formula C23H26N2O4 -+ xH2O
Linear Formula C23H26N2O4 -+ xH2O
MDL Number MFCD00149385
Quantity 100 g
Molecular Weight (g/mol) 394.46 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.