Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) selenide, 99%, Thermo Scientific™

Catalog No. AA1284009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1284009 10 g
AA1284018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1284009 Supplier Thermo Scientific Chemicals Supplier No. 1284009

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-00-0
Molecular Formula PbSe
Molecular Weight (g/mol) 286.20
MDL Number MFCD00016273
InChI Key GGYFMLJDMAMTAB-UHFFFAOYSA-N
Synonym lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g
PubChem CID 61550
IUPAC Name plumbaneselone
SMILES [Se]=[Pb]

Specifications

Melting Point 1,065°C
Physical Form Pieces
Quantity 10 g
UN Number UN3077
Formula Weight 286.15
Percent Purity 99%
Odor Product specific
Chemical Name or Material Lead(II) selenide
missing translation for 'healthHazard1' Danger
missing translation for 'healthHazard2' GHS H Statement
H300-H330-H360-H373
Fatal if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
missing translation for 'healthHazard3' GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.