Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead, Oil based standard solution, Specpure™ Pb 1000μg/g

Catalog No. AA1420818
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1420818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1420818 Supplier Thermo Scientific Chemicals Supplier No. 01420818
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134050

Specifications

Physical Form Liquid
Quantity 50 g
Grade Specpure
Chemical Name or Material Lead Oil based standard solution, Pb 1000μg/g
Hazard Category H304-H350-H360
Hazard Statement GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P281-P301+P310-P308+P313-P331-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.