Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Lead, Quant™ Test Strips

Catalog No. AA34933QA
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA34933QA 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA34933QA Supplier Thermo Scientific Chemicals Supplier No. 034933QA
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead, Test Strips
Quantity 1 Kit
MDL Number MFCD00147400
What concentrations of Pb (lead) can the Lead, Quant Test Strips test for?

The Lead, Quant Test Strips can measure concentrations of 20-500 ppm Pb+2 in aqueous solutions.

What is the protocol to use the Lead, Quant Test Strips?

We recommend the following protocol to use the Lead, Quant Test Strips:

  1. Take one strip from the container and immediately close the container.
  2. Dip the strip into the test solution to wet the reaction zone and remove excess liquid by shaking the test strip or drawing it across the edge of the vessel.
  3. Allow the reaction to proceed for ≤1 minute. Do not exceed 1 minute for the reaction time.
  4. Compare the color of the reaction zone on the strip with the color scale printed on the tube to determine the concentration of lead.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.