Learn More
Chemglass Life Sciences Chemglass lead ring, vinyl-coated C-shape - fits 125-500 mL flasks and beakers
SDP
Catalog No. NC3037304
Encompass
Vinyl-coated C-shape lead ring designed to stabilize and protect 125-500 mL flasks and beakers. The vinyl coating prevents marking or scratching of glassware and surfaces, while the C-shape allows slight manual adjustment for a snug fit; suitable for bench support or immersion in water baths.
  • Fits 125-500 mL flasks and beakers
  • Vinyl coating protects glassware and work surfaces
  • C-shape allows manual size adjustment for a snug fit
  • Made of lead for weight and stability
  • Suitable for bench support and immersion in water baths
Catalog No. NC3037304 Supplier Chemglass Life Sciences Supplier No. CLS4949001
Vinyl-coated C-shape lead ring designed to stabilize and protect 125-500 mL flasks and beakers. The vinyl coating prevents marking or scratching of glassware and surfaces, while the C-shape allows slight manual adjustment for a snug fit; suitable for bench support or immersion in water baths.
  • Fits 125-500 mL flasks and beakers
  • Vinyl coating protects glassware and work surfaces
  • C-shape allows manual size adjustment for a snug fit
  • Made of lead for weight and stability
  • Suitable for bench support and immersion in water baths