Learn More
Chemglass Life Sciences Lead Ring, Vinyl-Coated, C-Shaped - Fits 250 to 1,000 mL Flasks
SDP
Catalog No. NC3037329
Encompass
Vinyl-coated C-shaped lead ring designed to support and stabilize 250 to 1,000 mL laboratory flasks and vessels. The vinyl coating protects glassware and bench surfaces while the lead body provides weight for secure placement; the C-shape allows slight manual adjustment for a customized fit and facilitates immersion in water baths.
  • Fits 250 to 1,000 mL flasks and vessels.
  • Vinyl-coated finish prevents marking or scratching.
  • C-shaped design allows manual size variation for a customized fit.
  • Lead construction provides stable weighted support.
  • Inner diameter: 51 mm; outer diameter: 90 mm.
  • Weight: approximately 472 g.
Catalog No. NC3037329 Supplier Chemglass Life Sciences Supplier No. CLS4949002
Vinyl-coated C-shaped lead ring designed to support and stabilize 250 to 1,000 mL laboratory flasks and vessels. The vinyl coating protects glassware and bench surfaces while the lead body provides weight for secure placement; the C-shape allows slight manual adjustment for a customized fit and facilitates immersion in water baths.
  • Fits 250 to 1,000 mL flasks and vessels.
  • Vinyl-coated finish prevents marking or scratching.
  • C-shaped design allows manual size variation for a customized fit.
  • Lead construction provides stable weighted support.
  • Inner diameter: 51 mm; outer diameter: 90 mm.
  • Weight: approximately 472 g.