Learn More
SPEX CERTIPREP LLC 3P LEAD STANDARD 1000PPM 250ML
Catalog No. NC9842015
Encompass_Preferred
(HPS by Zeptometrix) Lead, Source: Pb Metal, Purity: 99.995%, Matrix: 2% HNO3
Catalog No. NC9842015 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000281250
(HPS by Zeptometrix) Lead, Source: Pb Metal, Purity: 99.995%, Matrix: 2% HNO3