Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Plastic bottle
10099-74-8,7697-37-2,7732-18-5
N2O6Pb
331.208
MFCD00134050
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
10099-74-8,7697-37-2,7732-18-5
N2O6Pb
331.208
MFCD00134050
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
For atomic adsorption spectroscopy
Concentration 997 to 1003μg/ml of Pb(2+) in 2 to 5% HNO3 (20°C)
Merck Index 15, 5469
Formula Weight 331.21
Physical Form Powder or Crystals
Grade AAS
Packaging Plastic bottle
Density 1.0200g/mL
Melting Point -3.0°C
Boiling Point 101.0°C
Color Brown to Red or Green
Quantity 100mL
Specific Gravity 1.02
Chemical Name or Material Lead standard solution, 1 mg/ml Pb in 2% HNO3
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement:
Keep only in original container.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger