Learn More
Cpi International LEAD STD 1000PPM 100ML
SDP
Supplier:  Cpi International S44001000281
Encompass
Single-Element Standards 1000microgram/mL, Element: Lead, Source: Pb metal, Purity: 99.999%, Matrix: 2% HNO3
Catalog No. NC9991827