Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Sulfide
SCI_ED

Catalog No. S75151 Shop All Fisherbrand Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S75151 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S75151

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black
CAS 1314-87-0
Physical Form Metallic Crystals, Powder Lump
Quantity 100 g
Molecular Formula PbS
Linear Formula PbS
MDL Number MFCD00016280
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES [S--].[Pb++]
IUPAC Name λ²-lead(2+) sulfanediide
Molecular Weight (g/mol) 239.30
PubChem CID 14819
Formula Weight 239.25
Chemical Name or Material Lead Sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.