Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Tetraacetate, 85% min., BAKER™, J.T. Baker™

Catalog No. 02003367 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-367 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-367 Supplier Avantor Supplier No. P36805

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 546-67-8
Molecular Formula C8H16O8Pb
Molecular Weight (g/mol) 447.408
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Quantity 100 g
Formula Weight 443.37
Percent Purity ≥85%
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.23kg
Chemical Name or Material Lead Tetraacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.