Learn More
Hach Company Lead TNTplus Vial Test (0.1-2.0 mg/L Pb), 25 Tests
Supplier:  Hach Company TNT850
Encompass
Lead TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.  For determination of Lead by the PAR Method with a working range of 0.1 - 2.0 mg/L Pb. Hach method 10216 has greatly simplified spectrophotometer based lead testing by eliminating the use of separatory glassware and organic solvents. Add Metals Prep Set (TNT890) to determine total lead (requires digestion).
Catalog No. 50-199-8141