Learn More
Sigma Aldrich Fine Chemicals Biosciences Lead wire, length 0.5 m, 0.6 mm diameter, high purity 99.999% | 7439-92-1 | MFCD00134050 | 1EA
Catalog No. 501771531
Encompass
Lead wire, length 0.5 m, 0.6 mm diameter, high purity 99.999% | Purity: 99.999% | Mol Wt: 207.2 | 7439-92-1 | MFCD00134050 | 1EA
This item is not returnable. View return policy
Catalog No. 50-177-1531 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF772172841EA
Lead wire, length 0.5 m, 0.6 mm diameter, high purity 99.999% | Purity: 99.999% | Mol Wt: 207.2 | 7439-92-1 | MFCD00134050 | 1EA