Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience LeadCare II Placement Program
GSA_VA

Catalog No. 23800100P Shop All Meridian Bioscience Products
Change view
Click to view available options
Sample Size:
0.05 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
23-800-100P 0.05 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-100P Supplier Meridian Bioscience Supplier No. 706760P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Point-of-care lead testing, cost-effective way to get started with no upfront investment in equipment

  • LeadCare™ II Placement Program - Unlock the benefits of point-of-care lead testing with the LeadCare™ II Placement Program - a simple, cost-effective way to get started with no upfront investment in equipment
  • With this annual agreement, your facility commits to purchasing 8 LeadCare™ II test kits per year (384 tests)
  • Your first order includes:
  • 4 LeadCare™ II test kits
  • 1 LeadCare™ II analyzer at no charge
  • Those initial 4 kits count toward your annual commitment
  • Meridian Bioscience retains ownership of the analyzer for the duration of the agreement and provides full warranty coverage as long as the agreement remains active, ensuring peace of mind and uninterrupted testing

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Point-of-Care Analyzer
Sample Type Capillary Whole Blood
For Use With (Application) Vitro Diagnostic
Analysis Time 3 min.
Sample Size 0.05 L
Calibration Electronic Calibration Button
Analytical Method Anodic Stripping Voltammetry (ASV)
Cuvettes Yes
Warranty Warranty Included
Voltage 100/240 V
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.