Learn More
Enzo Life Sciences LEADING LIGHT Wnt Cell Line Medium Pack (1 Pack)
SDP
Catalog No. NC0652705
Encompass_Preferred
For use with the LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway..The LEADING LIGHT.60 mL, Cell Growth Medium Concentrate.4.5 mL, Cell Assay Medium Concentrate.10 mL, Cell Freezing Medium..Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter medium. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C
Catalog No. NC0652705 Supplier Enzo Life Sciences Supplier No. ENZ600030001
For use with the LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway..The LEADING LIGHT.60 mL, Cell Growth Medium Concentrate.4.5 mL, Cell Assay Medium Concentrate.10 mL, Cell Freezing Medium..Note: the complete LEADING LIGHT Wnt Reporter Assay Starter Kit (Prod. No. ENZ-61001) is available. Alternative name: Wnt reporter medium. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C