Learn More
Enzo Life Sciences LEADING LIGHT Wnt Reporter Assay Starter Kit (1x96wells)
SDP
Catalog No. NC0864692
Encompass_Preferred
Chemiluminescent cell-based assay for discovering Wnt pathway modulators. The LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway. Alternative Name: Wnt reporter assay. Applications: FUNC. Chemiluminescence, HTS. Contents: LEADING LIGHT Wnt Reporter Cell Line, Cell Assay and Growth Medium Concentrate, Cell Freezing Medium, Luciferase Substrate and Assay buffer, HEPES, Wnt3a Protein (positive control), and Dkk-1 Protein
Catalog No. NC0864692 Supplier Enzo Life Sciences Supplier No. ENZ610010001
Chemiluminescent cell-based assay for discovering Wnt pathway modulators. The LEADING LIGHT. .The assay has been used successfully in different assay formats (including HTS applications) to identify several distinct categories of small molecule compounds that modulate the Wnt signaling pathway. Alternative Name: Wnt reporter assay. Applications: FUNC. Chemiluminescence, HTS. Contents: LEADING LIGHT Wnt Reporter Cell Line, Cell Assay and Growth Medium Concentrate, Cell Freezing Medium, Luciferase Substrate and Assay buffer, HEPES, Wnt3a Protein (positive control), and Dkk-1 Protein